Imprimer
Self contained biological indicators (SCBI)
Fournisseur: CROSSTEX
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SCS-06 SCS-05
NAMSSCS-06EA 670 EUR
NAMSSCS-06 NAMSSCS-05
Self contained biological indicators (SCBI)
Indicateurs de stérilisation
Self-contained biological indicators (SCBIs) for monitoring steam sterilisation consist of a durable PC vial with cap, a crushable glass ampoule with Trypticase® soy broth (TSB) modified with pH indicator, and a disc inoculated with Geobacillus stearothermophilus spores.
  • Reduced incubation time of 24 hours
  • Visually distinct positive results indicated by the media transitioning from purple to yellow
  • Chemical indicator on label provides instant indication of exposure
Storage condition: 15 to 30 °C
Relative humidity: 20 – 70%
Certifications: Complies to ISO 11138-1, ISO 11138-3, USP.
Attention: For industrial use only.