Imprimer
Cooling thermostats, PRO Series
Fournisseur: LAUDA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L000112 L000114 L000007 L000008 L000009 L000010 L000011 L000012 L000013 L000014 L000015 L000016 L000017 L000018 L000090 L000092 L000094 L000096 L000098 L000100 L000110 L000116 L000118 L000120 L000170 L000172 L000174 L000176 L000178 L000180 L000190 L000192 L000194 L000196 L000198 L000200
LAUDL000112EA 12020 EUR
LAUDL000112 LAUDL000114 LAUDL000007 LAUDL000008 LAUDL000009 LAUDL000010 LAUDL000011 LAUDL000012 LAUDL000013 LAUDL000014 LAUDL000015 LAUDL000016 LAUDL000017 LAUDL000018 LAUDL000090 LAUDL000092 LAUDL000094 LAUDL000096 LAUDL000098 LAUDL000100 LAUDL000110 LAUDL000116 LAUDL000118 LAUDL000120 LAUDL000170 LAUDL000172 LAUDL000174 LAUDL000176 LAUDL000178 LAUDL000180 LAUDL000190 LAUDL000192 LAUDL000194 LAUDL000196 LAUDL000198 LAUDL000200
Cooling thermostats, PRO Series
Thermostats à circulation Systèmes à circulation fermée Circulateurs de refroidissement
The PRO cooling bath thermostats for internal bath applications are equipped with a multi-stage adjustable pump. This ensures good mixing of the bath. It has 360º turnable and detachable control panel for remote control.
  • Touchscreen can be operated with gloves
  • Internal Varioflex pump (pressure-suction pump) with eight pump levels
  • Programmer with one program, 20 segments whereas 100 programs, with 50 segments for command unit
  • Individual access rights for up to 20 users
  • Enables ramp function for easy computation
  • Hybrid cooling for cooling the refrigeration machine with ambient air or additional cooling water
High contrast OLED display for units with Base control, whereas large colour touchscreen (145 mm) with multi touch operation for units with Command Touch, with timer and count down display.
Max. flow rate / max. pressure:
Pump pressure/suction: 0,7/0,4 bar
Pump flow pressure/suction: 22/20 L/min
Informations de livraison: Supplied with bath covers and 2 hose olives with cap nuts for the cooling coil.