Imprimer
Heating thermostats with stainless steel bath, PRO series
Fournisseur: LAUDA
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
L000001 L000002 L000003 L000004 L000005 L000006 L000106 L000107 L000108 L000126 L000127 L000128 L000186 L000187 L000188 L000206 L000207 L000208
LAUDL000001EA 6460 EUR
LAUDL000001 LAUDL000002 LAUDL000003 LAUDL000004 LAUDL000005 LAUDL000006 LAUDL000106 LAUDL000107 LAUDL000108 LAUDL000126 LAUDL000127 LAUDL000128 LAUDL000186 LAUDL000187 LAUDL000188 LAUDL000206 LAUDL000207 LAUDL000208
Heating thermostats with stainless steel bath, PRO series
Thermostats à circulation Systèmes à circulation fermée Thermostats chauffants
The PRO heating bath thermostats are optimised for internal bath applications and offer excellent temperature stability. The removable operating units, including mount, allow considerable reduction in the height of the device.
  • Touchscreen can be operated with gloves
  • Internal Varioflex pump (pressure-suction pump) with eight pump levels
  • Programmer with one program, 20 segments or 100 programs, with 50 segments for command unit
  • Individual access rights for upto 20 users
  • Enables ramp function for easy computation
Units have a 360º turnable and detachable control panel for remote control. High contrast OLED display for units with Base control, whereas large colour touchscreen (145 mm) with multi touch operation for units with Command Touch, with timer and count down display.
Max. flow rate / max. pressure:
Pump pressure/suction: 0,7/0,4 bar
Pump flow pressure/suction: 22/20 L/min
Informations de livraison: Supplied with bath covers and 2 hose olives with cap nuts for the cooling coil.