Imprimer
Micro-Tube homogeniser system, Bel-Art ProCulture, SP Bel-Art
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F650009001 F650000002 F650009002 F650000005 F650000006 F650009006 F651000000
BELAF650009001EA 424 EUR
BELAF650009001 BELAF650000002 BELAF650009002 BELAF650000005 BELAF650000006 BELAF650009006 BELAF651000000
Micro-Tube homogeniser system, Bel-Art ProCulture, SP Bel-Art
Homogénéisateurs Homogénéisateurs autonomes
Hand-held, cordless homogeniser features a simple push in / pull out pestle exchange and is ideal for thorough and faster processing of one to many samples.
  • Pestles and tubes assure tight tolerances and thorough homogenisation and grinding of materials resulting in optimal efficiency of pellet reconstitution or re-suspension
  • Pestles and tubes are chemical and heat resistant; colorant-free to eliminate potential leaching of heavy metals into samples
  • Tubes have 25% thicker bottoms than industry standard to ensure integrity during use
  • Each tube has frosted side and cap surfaces for labeling, a positive seal 'snap-cap' design and a pierceable top for optimal functionality
  • Pestles have a unique ribbed shaft that provides precise finger control of homogenisation for shear sensitive samples; also fit most motorised mixer adapters
Matched pestle tips and micro tubes can be used either with the homogeniser or with hand processing.
Certifications: Manufactured and packaged in a class 10,000 cleanroom; pestles and tubes are certified DNase, RNase and Pyrogen-free, and made of animal and phthalate/phthalate-ester free polypropylene with lot control and material traceability.
Informations de livraison: Homogeniser includes cordless motor, 2 AA batteries and pestle adapter.
Cdt: Available in individually wrapped tubes and pestles over packed in resealable bags of 100 each or bulk packed in resealable bag with an additional heat-seal above the standard closure that indicates if the package has been opened.
Attention: Sterilised products are not returnable.