Imprimer
HyClone™ CDM4CHO, CHO Cell Culture Platform Liquid Media
Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30557.02 SH30556.01 SH30556.03 SH30557.LS SH30558.02
SH30557.02EA 124 EUR
SH30557.02 SH30556.01 HYCLSH30556.03 HYCLSH30557.LS SH30558.02
HyClone™ CDM4CHO, CHO Cell Culture Platform Liquid Media
Milieux de culture cellulaire
HyQ® CDM4CHO™ is a chemically-defined protein-free medium containing no animal derived components. It is designed to increase the process yields for the industrial manufacture of recombinant proteins using a variety of CHO cell clones. HyQ® CDM4CHO™ has been successfully tested in various culture systems, including T-flasks, shaker flasks and bioreactors. HyQ® CDM4CHO™ contains Pluronic® F68 and L-glutamine, and does not contain phenol red.
  • Metabolically designed for high cell yield and recombinant protein production
  • Allows for direct or sequential adaptation
  • Designed for large-scale culture applications, including perfusion and fed-batch strategies
  • All components fully traceable
  • Manufactured under cGMP
HyClone CDM4CHO contains Pluronic F68 and L-glutamine, and does not contain phenol red. It is also available without L-glutamine to support the GS gene expression system.