Imprimer
Tamper evident closures for PET/PETG square media bottles, HDPE
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
NALG342141-0384EA 171 EUR
NALG342141-0384
Tamper evident closures for PET/PETG square media bottles, HDPE
Capuchons Bouchons pour flacons
Closure, tamper evident, specially designed for use with PET/PETG square media bottles.
  • Natural HDPE closures are ADCF (Animal Derived Component-Free)
  • Fits Nalgene PET and PETG Media Bottles with 38-430 neck finish and leakproof guarantee
  • Easy application and breaking of the tamper evident seal ring
  • Gamma irradiated to provide 10⁻⁶ Sterility Assurance Level (SAL)
  • Flat top surface for possible printing or labelling
Cdt: Closures are sterile and bulk packed.