PathoGreen™ Histofluorescent Stain 1000X water
Marques : BiotiumTotal Evaluation: 0
Moyenne des Notes : 0.0 sur 5
PathoGreen histofluorescent stain is an anionic green fluorescent dye functionality similar to Fluoro-Jade dyes.
- Ex/Em 497/520 nm
- Dyes stain degenerating neurons and their processes
- Anionic green fluorescent dye functionality
These dyes stain degenerating neurons and their processes after exposure to neurotoxic insults in brain sections and cultured neurons.
The mechanism of neuronal staining by anionic fluorescent dyes has not been determined. It has been proposed that the negatively charged dyes bind to positively charged polyamines or other molecules specifically generated in dying neurons.
Fluoro-Jade is a registered trademark of Histo-Chem, Inc.