Imprimer
Baths and circulators, stainless steel, SAHARA series
Fournisseur: THERMO ELECTRON LED
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
1541131 152-1491 152-1301 152-1211 153-1211 153-1131 153-1071 153-1031 1541071 1541491 156-1071 1561451 1551451 1521451 1531451
HAKA1541131EA 5270 EUR
HAKA1541131 HAKA152-1491 HAKA152-1301 HAKA152-1211 HAKA153-1211 HAKA153-1131 HAKA153-1071 HAKA153-1031 HAKA1541071 HAKA1541491 HAKA156-1071 HAKA1561451 HAKA1551451 HAKA1521451 HAKA1531451
Baths and circulators, stainless steel, SAHARA series
Thermostats à circulation Systèmes à circulation ouverte Bains avec circulateurs détachables
The SAHARA series are suitable for internal or external circulation and have a choice of eight different controllers that are suitable for various different applications.
  • All thermostats feature an intuitive, user interface with bright display to view important readings
  • Thermostat can be turned around and positioned on all four sides on the bath for easy viewing
  • Each system comes with a quick-start guide for simple setup and operation
  • Save up to 80% of energy costs and thousands of kilowatt hours during the product lifetime
Certifications: Units are RoHS-compliant (lead- and mercury-free) and UL/CSA compliant, ensuring safe operation in the unlikely event of a fault.
Informations de livraison: Supplied with communication cables, bridge with gasket and thumbscrews, fittings, clamps, pump plug for external circulation and power cord.