Imprimer
Bottle, narrow neck, round, HDPE, with closure, Nalgene®
Fournisseur: EP SCIENTIFIC
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
156-125W/N/LP 150-01W/N/LPM 150-01W/N/LP 156-125W/N/LPM 157-250W/N/LP 157-250W/N/LPM
EPSC156-125W/N/LPEA 1010 EUR
EPSC156-125W/N/LP EPSC150-01W/N/LPM EPSC150-01W/N/LP EPSC156-125W/N/LPM EPSC157-250W/N/LP EPSC157-250W/N/LPM
Bottle, narrow neck, round, HDPE, with closure, Nalgene®
Bottles Plastic Bottles
Particulate-cleaned bottles used for storage of the product or high purity samples.
  • Reliable and consistent quality
  • Bulk intermediate and storage in cleanroom or controlled environment use
  • Pharmaceutical and biotech use
Bottles are double-bagged and ready to take into your cleanroom with no extra preparation. Low metals certification includes aluminum, calcium, copper, iron, potassium, magnesium, manganese, sodium and zinc at <10 ppb.
Informations de livraison: Suppliied with the certificate of analysis.
Attention: *The product(s) on this page are outside the scope of the EU Directive 2019/904