Imprimer
Cooler, ice-free, Corning® XT Starter
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
432014 432015
479-2177EA 271 EUR
479-2177 CORN432015
Cooler, ice-free, Corning® XT Starter
Coolers Benchtop Coolers
An open-platform ice-free cooler that accommodates most CoolRack® and CoolSink® modules.
  • Consists of a base, cooling core and a lid
  • Includes a 1 to 8 °C temperature indicator for visual assurance of proper temperature
  • To use with most Corning CoolRack® and CoolSink® Modules
  • The internal cooling or freezing cores provide 0,5 to 4 °C for up to 4 hours
  • One module capacity
  • Purple color
Low profile and small footprint make it ideal for use in the hood, keeping samples cold (0,5 to 4,0 °C) up to four hours. 1 to 8 °C temperature indicator provides visual assurance of temperature performance. To extend the cooling duration, keep an additional XT cooling core in the freezer and rotate the cores as needed.