Imprimer
Sampling bag, sterile, for medical and food applications, Intervoid®
Fournisseur: COVERIS
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
COV-112S COV-113S COV-114S COV-115S COV-116S COV-117S COV-118S COV-119S COV-120S COV-121S
129-1163EA 210 EUR
129-1163 129-1164 129-1165 129-1166 129-1167 129-1168 129-1169 129-1170 129-1171 129-1172
Sampling bag, sterile, for medical and food applications, Intervoid®
Sachets Sachets d'échantillonnage
The Intervoid® secure sampling bags are designed to protect and transport mainly critical samples ; they combine the highest levels of tamper evident security and tracking technologies to bring real benefits to multiple analytical processes.
  • High premium sterilization standards (10-6 gamma): bags closed on production, can be open by pre-cut under the security tape ensuring only the bag being used is exposed, greatly reducing wastage
  • Can be used both for primary or secondary packaging
  • Leak resistant
  • Single trip by design to avoid any risk of cross contamination or sample confusion
  • Traceability: barcode, sequential numbering allowing, detachable receipt
  • High-security: closure, unique marking system (pat. Pend.), micro printing
Note: The dimensions (L×W) given in the table are internal dimensions.
Certifications: Approved food contact.
Cdt: The Sterile Secure MEDICAL Sampling Bag are packed in inner bags of 10.
The Sterile Secure FOOD Sampling Bag are packed in inner bags of 25.