Imprimer
Screw cap trays for 2D barcoded storage tubes
Fournisseur: THERMO MATRIX TECHNOLOGIES
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
4906 4477 4477BLU 4477GRE 4477GRY 4477PIN 4477PUR 4477RED 4477WHI 4477YEL
479-2066EA 81.5 EUR
479-2066 479-2052 479-2053 479-2054 479-2055 479-2056 479-2057 479-2058 479-2059 479-2060
Screw cap trays for 2D barcoded storage tubes
Plateaux/Récipients/Pots Plateaux et récipients de laboratoire
Simplify sample storage with these color-coded caps and inserts.
Superior cap design
Gasketed caps ensure critical sample protection over the storage lifetime
Soft gasket compresses to ensure a proper seal, preventing contamination from outside the tube
Cap material: Virgin class VI medical grade polypropylene with virgin class VI medical grade silicone O-ring
ScrewTop caps are compatible with storage down to −196 ºC
Cap options simplify sample identification
Cap trays feature standard microplate format compatible with automated equipment
Caps are available in seven color options to enable sample identification
Color inserts snap into cap for a secure fit; inserts are removable
Inserts are available in six colors
All caps and inserts are supplied free from DNA, Rnase, Dnase, endotoxins and cytotoxins.