Imprimer
CO₂ absorbent, 20 to 30 Mesh, Capacity: 40 to 50%, Product of Arthur H. Thomas Company, Ascarite(II)®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
208095000 208090025 208091000
ACRO208095000EA 582 EUR
ACRO208095000 ACRO208090025 ACRO208091000
CO₂ absorbent, 20 to 30 Mesh, Capacity: 40 to 50%, Product of Arthur H. Thomas Company, Ascarite(II)®
Desiccators Desiccants
CAS No.: 81133-20-2
MDL No.: MFCD00132264
UN: 1823
Hazard Class: 8
Packing group: II
GHS Symbols: GHS05
Appearance: Beige to light brown Granules
Storage Temperature: Ambient
Signal word: Danger