Imprimer
CO₂ absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Ascarite(II)®
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
208085000 208080025 208081000
ACRO208085000EA 540 EUR
ACRO208085000 ACRO208080025 ACRO208081000
CO₂ absorbent, 8 to 20 mesh, product of Arthur H. Thomas Company, Ascarite(II)®
Desiccators Desiccants
CAS No.: 81133-20-2
MDL No.: MFCD00132264
UN: 1823
Hazard Class: 8
Packing group: II
GHS Symbols: GHS05
Appearance: Brown Granules
Storage Temperature: Ambient
Signal word: Danger