Imprimer
Beta-galactosidase substrates
Marques : Biotium
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
10015 10014 10012 10013 10024
10015.EA 242 EUR
10015. 10014. 10012. 10013. 10024.
Beta-galactosidase substrates
Colorants Colorants d'imagerie in vivo
Chromogenic and fluorogenic beta-galactosidase substrates.
White solid form soluble in water and DMSO.
Informations de commande: Includes 5 mg of X-gal, red-beta-D-gal, rose-beta-D-gal, purple-beta-D-gal and green-beta-D-gal.
Attention: Store at −20 °C and protect from light.
https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=d568646624b7b6dc9e28e334c4beecd462a1351c&vid=d568646624b7b6dc9e28e334c4beecd462a1351c&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=5c5c39c007d3027b76b2616aa69f15bc9b4ce194&vid=5c5c39c007d3027b76b2616aa69f15bc9b4ce194&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=6cbad528fc0a061e68483882cd9ac93df3f0e675&vid=6cbad528fc0a061e68483882cd9ac93df3f0e675&prid=web&clid=SAPDAM https://digitalassets.avantorsciences.com/adaptivemedia/rendition?id=fd27bd3a9697599204412f7eb5d1f59e7b9d41af&vid=fd27bd3a9697599204412f7eb5d1f59e7b9d41af&prid=web&clid=SAPDAM