Imprimer

Anti-IgG Llama Polyclonal Antibody (CF™ 488A)

Fournisseur: Biotium
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

20449-50UL 20449-500UL
20449-50ULEA 127 EUR
20449-50UL 20449-500UL
Anti-IgG Llama Polyclonal Antibody (CF™ 488A)
Anticorps
Llama anti-rabbit CF™ dye conjugates are prepared by labeling affinity purified llama anti-rabbit IgG (H+L) with a selection of CF™ dyes. CF™ dyes offer combined advantages in brightness, photostability, and specificity compared other fluorescent dyes. CF™488A (Ex/Em 490/515 nm) is a superior alternative to the traditional green fluorescent dye FITC (or fluorescein) owing to its exceptional brightness and photostability. CF™488A has brightness and photostability comparable to Alexa Fluor® 488.

Alexa Fluor® is a registered trademark of Thermo Fisher Scientific.

Type: Secondary
Antigen: IgG
Clonality: Polyclonal
Clone:
Conjugation: CF488A
Epitope:
Host: Llama
Isotype:
Reactivity: Rabbit
Commander maintenant

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR