Imprimer
Protein IgG Plus orientation kits, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44893 44990
PIER44893EA 804 EUR
PIER44893 PIER44990
Protein IgG Plus orientation kits, Pierce™
Protéines purification Protein Purification Kits
Pierce™ Protein IgG Plus Orientation Kits uses a crosslinker to covalently attach antibody that is bound to Protein A (or Protein G) agarose resin, thereby creating a reusable affinity column for purification of antigens.
  • Contaminant free antigen purification and less antibody leaching
  • Covalent, leak resistant antibody immobilisation
  • Gentle binding and crosslinking conditions
  • Less antibody leaching
  • Crosslinked 6% beaded agarose with recombinant Protein A (or Protein G) thus making it high quality resin
  • Oriented immobilisation that is able to capture passing antigen in the mobile phase without steric hindrance
Informations de livraison: Kit includes Recombinant Protein A [or Protein G] columns, 2 ml (2 columns), disuccinimidyl suberate [DSS] (2×13 mg), PBS pack (makes 500 ml), blocking buffer (6 ml), IgG elution buffer (15 ml), antibody binding/wash buffer (120 ml), and column accessories.