Imprimer
Mouse IgG1 Fab and F(ab')2 preparation kits, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
44980 44680
PIER44980EA 880 EUR
PIER44980 PIER44680
Mouse IgG1 Fab and F(ab')2 preparation kits, Pierce™
Protéines purification Protein Purification Kits
The Pierce™ Mouse IgG1 Fab and F(ab')2 Preparation Kits use immobilised ficin protease to efficiently digest mouse IgG1 into Fab or F(ab')2 fragments.
  • Kit procedure optimised for mouse IgG1
  • Kit includes all reagents needed to prepare and purify antibody fragments
  • Immobilised ficin (beaded agarose resin) provides for control of the digestion reaction and complete removal of resulting antibody fragments from the proteolytic enzyme
Informations de livraison: Kit includes ficin agarose (2,5 ml), cysteine-HCl (1 g), mouse IgG1 digestion buffer (120 ml), NAb™ Protein A Plus spin column (1×1 ml), Protein A binding buffer (120 ml), IgG elution buffer (120 ml), Zeba desalt spin columns, 7K MWCO (10×2 ml), spin columns (10×0,8 ml) and microcentrifuge tubes(30×2 ml).