Imprimer
Trypsin protease, MS grade
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
90057 90058 90059 90305
PIER90057EA 103 EUR
PIER90057 PIER90058 PIER90059 PIER90305
Trypsin protease, MS grade
Protéines extraction Réactifs pour l'extraction des protéines
Pierce™ Trypsin Protease, MS Grade, is highly purified porcine trypsin that has been chemically modified for maximum activity and stability in proteomic applications.
  • Cleaves at the carboxyl side of lysine and arginine residues with greater than 95% specificity
  • No detectable chymotrypsin activity thus making it highly pure
  • Chemically modified for reduced autolytic activity
Applications include in-gel digestion of proteins from 1-D or 2-D gels and in-solution tryptic digestion of proteins. The selectivity of this enzyme is critical for reproducible protein digestion and mass spectrometry-based protein identification. It has been treated with TPCK to eliminate chymotryptic activity, improving digestion specificity. The enzyme is most active in pH ranges 7 to 9 and can be reversibly inactivated at pH <4.