Imprimer
Protein L agarose, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20510 20512
PIER20510EA 432 EUR
PIER20510 PIER20512
Protein L agarose, Pierce™
Protéines purification Protein Purification Resins
Pierce™ Protein L agarose is a premium quality Protein L beaded agarose resin for use in a variety of antibody affinity purification methods.
  • Immobilised Protein L is ideal for selective purification of human and mouse antibodies that have kappa light chains
  • Support is crosslinked 6% beaded agarose (CL-6B), the most popular resin for protein affinity purification methods
  • Protein L is immobilised by charge-free, leach-resistant covalent bonds, resulting in low non specific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilised Protein L
Binding capacity of 5 to 10 mg human IgG/ml resin