Imprimer
Mass spectrometry reagents, MALDI matrices, single-use
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
90032 90033 90035 90031
PIER90032EA 276 EUR
PIER90032 PIER90033 PIER90035 PIER90031
Mass spectrometry reagents, MALDI matrices, single-use
Protéines purification Réactifs de purification des protéines
MALDI Matrices are highly purified, recrystallised reagents packaged in convenient single-use tubes sufficient for analysing up to 96 samples.
  • Single-use packaging saves time and reduces waste
  • Provides accurate quantity of matrix every time
  • Readily automated
  • Highly sensitive
Matrix assisted laser desorption ionisation (MALDI) matrices produce the cleanest mass spectrometric (MS) spectra when they are recrystallised and prepared daily, which is inconvenient and typically results in significant waste. The packaging of purified, recrystallised alpha-cyano-4-hydroxy-cinnamic acid (CHCA), sinapinic acid (SA) and 2,5-dihydroxybenzoic acid (DHB) MALDI matrices makes it easy to prepare high quality MALDI reagents in minutes.