Imprimer
Protein G agarose, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20398 20399 20397
PIER20398EA 404 EUR
PIER20398 PIER20399 PIER20397
Protein G agarose, Pierce™
Protéines purification Protein Purification Resins
Pierce™ Protein G agarose is a premium quality affinity resin for antibody purification.
  • Ideal for polyclonal IgG purification from mouse, human, cow, goat, and sheep serum, including human IgG3 and mouse IgG1 isotypes
  • Agarose resin support is crosslinked 6% beaded agarose (CL-6B)
  • Protein G made by charge-free, leach-resistant covalent bonds, resulting in low nonspecific binding and enabling multiple uses without decline in yield
  • Standard capacity beads with a normal load of immobilised Protein G
Binding capacity: 11 to 15 mg human IgG/ml resin