Imprimer
Streptavidin HRP conjugates, high sensitivity, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21130 21132 21134
PIER21130EA 292 EUR
PIER21130 PIER21132 PIER21134
Streptavidin HRP conjugates, high sensitivity, Pierce™
Protein Purification Protein Purification Reagents
Pierce™ High Sensitivity Streptavidin-HRP is a peroxidase-conjugated streptavidin biotin-binding protein that provides signal amplification like poly-HRP and exceptional storage stability.
  • Detect low levels of target without background; obtain high signal-to-noise ratios
  • Less conjugate enough in Western blotting and ELISA applications to obtain excellent results
  • Compatible with typical chemiluminescent, fluorescent and colorimetric peroxidase substrates
  • Ready-to-use stabilised liquid format means there is no waiting for thawing and no need to aliquot