Imprimer
Pierce™ HisPur™ Cobalt Agarose Spin Kits for Antibody Purification
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
89964 89965 89966 89967 89968 89969 90090 90091 90092
PIER89964EA 148 EUR
PIER89964 PIER89965 PIER89966 PIER89967 PIER89968 PIER89969 PIER90090 PIER90091 PIER90092
Pierce™ HisPur™ Cobalt Agarose Spin Kits for Antibody Purification
Protéines purification Protein Purification Kits
Pierce™ HisPur™ Cobalt Purification Kit is designed for the efficient purification of recombinant polyhistidine-tagged proteins from bacterial, mammalian and baculovirus-infected cells.
  • Obtain <10 mg of pure His-tagged protein per ml of resin
  • Resin can be reused several times or discarded after single use
  • Cobalt-chelate coordination core binds fewer host protein contaminants
  • Purify proteins under native or denaturing conditions
His-tagged proteins are purified from total soluble protein extract using a cobalt-charged tetradentate chelator immobilised onto 6% crosslinked agarose. Many immobilised metal affinity chromatography (IMAC) resins contain nickel (Ni²⁺) for purifying His-tagged proteins. Although Ni²⁺ chelate resins achieve high protein yields, purity is often sub-optimal, requiring additional clean-up steps. Cobalt achieves both high yield and purity with minimal optimisation. The HisPur™ Cobalt Resin binds fewer non specific proteins, displays less metal leaching and enables less stringent elution conditions compared to Ni²⁺ resins.
Informations de livraison: Complete kit contains HisPur™ Cobalt spin columns, pre-formulated Equilibration/Wash and Elution Buffers, collection tubes and tip plugs. Resin and spin columns are also available separately.