Imprimer
Pierce™, UltraLink™ Affinity Chromatography Resin, Immobilised Protein A/G
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
53132 53133 53135
PIER53132EA 477 EUR
PIER53132 PIER53133 PIER53135
Pierce™, UltraLink™ Affinity Chromatography Resin, Immobilised Protein A/G
Protéines purification Protein Purification Resins
Pierce™ Protein A/G UltraLink™ and Protein A/G Plus UltraLink™ resins are extremely durable beaded polyacrylamide supports to which recombinant Protein A/G has been immobilised for use in IgG purification methods. Immoblised proteins are covalently coupled to an agarose support to provide an efficient method of purifying antibodies.
  • Specifically binds IgG from several mammalian species, especially most subtypes of human and rabbit IgG
  • Acrylamide-based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
  • Used to bind and purify IgG in gravity-flow, spin-column or medium-pressure chromatography methods
Protein A/G UltraLink™ resin binding capacity: >20 mg human IgG/ml resin
Protein A/G Plus UltraLink™ resin binding capacity: >28 mg human IgG/ml resin