Imprimer
Donor horse serum, US orgin, HyClone™
Fournisseur: HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30074.02 SH30074.03HI SH30074.02HI
HYCLSH30074.02EA 17 EUR
HYCLSH30074.02 HYCLSH30074.03HI HYCLSH30074.02HI
Donor horse serum, US orgin, HyClone™
Sérums
HyClone™ donor horse serum, designed to supplement cell growth, is collected from selected donor herds that receive regular veterinary inspection and care.
  • Horses are bled in an enclosed facility that is specially equipped for aseptic collection
  • Coggins tests for equine infectious anemia are routinely performed on each horse
  • Serum filtered using sequential 100 nm pore-size rated filters
Supports hybridoma and antibody production. Meets specifications of \<=10EU/mL of endotoxin and \<=10mg/dL of hemoglobin. Filtered through triple 100nm pore-sizeEn savoir plus