Imprimer
Affinity coupling resins, AminoLink™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20381 20382
PIER20381EA 190 EUR
PIER20381 PIER20382
Affinity coupling resins, AminoLink™
Protéines purification Protein Purification Resins
AminoLink™ coupling resin is crosslinked, 4% beaded agarose that has been activated with aldehyde groups to enable covalent immobilisation of antibodies and other proteins through primary amines.
  • Ideal for antibodies and other proteins – immobilise molecules via primary amines (-NH2)
  • Efficient (>85%) coupling over a wide range of pH (4 to 10) and buffer conditions (PBS or other non-amine buffer, with or without organic solvent)
  • Coupling reaction results in stable, leak-resistant secondary amine bond between resin and ligand
  • Better than immobilisation to CNBr-activated agarose as the bond is more stable and uncharged, resulting in less non specific binding in affinity purification procedures
  • Prepared affinity resin is adaptable to column and batch affinity techniques and the resin is reusable for typical applications based on protein binding interactions