Imprimer
HyClone™ CDM4HEK293
Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30858.02 SH30859.01
SH30858.02EA 115 EUR
SH30858.02 HYCLSH30859.01
HyClone™ CDM4HEK293
Milieux de culture cellulaire
HyClone™ CDM4HEK293 media are chemically-defined, animal-derived component free (ADCF) cell culture media.
  • Allows for direct or sequential adaptation
  • Component traceability
  • Manufactured under cGMP
  • Metabolically designed for high cell yield and recombinant protein/vector production
HyClone CDM4HEK293 is a regulatory-friendly medium developed through the Metabolic Pathway Design to support the extremely high productivity and cell density in suspension cultures.