Imprimer
Agarose I, Pierce™
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
17850 17852
PIER17850EA 353 EUR
PIER17850 PIER17852
Agarose I, Pierce™
Gels d'électrophorèse Agarose et acrylamide
Agarose I (molecular biology grade) is a highly purified and carefully blended formulation that provides uniform lot-to-lot consistency for preparation of electrophoresis gels.
  • Standard melting temperature agarose for electrophoresis
  • Separation range: 100 bp to >30 kb