Imprimer
Monomeric avidin agarose, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20228 20267
PIER20228EA 616 EUR
PIER20228 PIER20267
Monomeric avidin agarose, Pierce™
Protéines purification Protein Purification Resins
Pierce™ Monomeric avidin agarose is immobilised onto 4% beaded agarose for purifying biotinylated proteins, peptides and other molecules.
  • Non-denaturing purifies biotinylated products using mild elution conditions (2 mM free biotin)
  • Reusable at least 10 times, with marginal loss in biotin binding capacity (approximately 2,5% decrease per regeneration)
  • Retains biotin-binding specificity and low non specific binding of native avidin
  • Binding capacity is >1,2 mg biotinylated BSA/ml resin