Imprimer
Pierce™ Avidin Agarose
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
20219 20225
PIER20219EA 298 EUR
PIER20219 PIER20225
Pierce™ Avidin Agarose
Protéines purification Protein Purification Resins
Pierce™ Avidin agarose is purified avidin immobilised on crosslinked 6% beaded agarose for the affinity purification of biotinylated macromolecules.
  • Strong, nearly irreversible biotin:avidin interaction
  • Approximate binding capacity >20 µg biotin/ml resin