Imprimer
Calf serum, iron supplemented, US origin, HyClone™
Fournisseur: HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30072.02 SH30072.04 SH30072.02HI SH30072.03HI SH30072.04HI SH30072.03IR25 SH30072.04IH25 SH30072.04IR25 SH30072.04IR30
HYCLSH30072.02EA 20.7 EUR
HYCLSH30072.02 HYCLSH30072.04 HYCLSH30072.02HI HYCLSH30072.03HI HYCLSH30072.04HI SH30072.03IR25 SH30072.04IH25 SH30072.04IR25 SH30072.04IR30
Calf serum, iron supplemented, US origin, HyClone™
Sérums
HyClone™ iron-supplemented bovine calf serum contains additional iron to boost serum transferrin levels to physiological levels.
  • Sourced in the USA with complete traceability back to original source
  • Cost-effective alternative to FBS
  • Age of animal at time of collection approximately 16 to 24 weeks
  • Low in antibodies and high in growth factors
  • Virus panel testing according to 9 CFR 113.53
  • Supplemented with iron and growth-promoting factors
HyClone Iron-Supplemented Bovine Calf Serum, U.S. origin is collected by venipuncture from formula-fed veal calves,En savoir plus