Imprimer
ESI positive ion calibration solution, LTQ, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
88322 88323
PIER88322EA 301 EUR
PIER88322 PIER88323
ESI positive ion calibration solution, LTQ, Pierce™
Étalons Étalons pour colorimétrie et spectroscopie
Positive ion calibration solutions are mixtures of highly purified ionisable molecules specifically designed for positive mode calibration of ion trap and orbitrap instruments.
  • Mass spectrometry grade reagents in a non-leachable container
  • Load the mass reference standard into a syringe and inject into the instrument
It is a ready-to-use liquid formulation ideal for quickly performing the routine calibration required to maintain the robust performance of Thermo Scientific mass spectrometers.