Imprimer
SP Bel-Art View Pack™ microscope slide holders
Fournisseur: Bel-Art Products, a Part of SP
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
F441700000 F441710000
467-0136EA 340 EUR
467-0136 467-0137
SP Bel-Art View Pack™ microscope slide holders
Porte-lames
Microscope slide holders, with ring binder.
  • Standard 230×300 mm three-ring binder comes complete with ten 220×270 mm vinyl slide holder pages, enough for 160 slides
  • Each page can hold 16 slides, in individual pockets with a white back and clear front
  • Center flap keeps slides from falling out
  • Fold the page at the center crease to retrieve slides
Refill View-Pack™ microscope slide holder pages – Pack of ten are available.