Imprimer
Dulbecco's phosphate buffered saline (DPBS), ES cell qualified, HyClone™ AdvanceSTEM™
Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30850.02 SH30850.03
HYCLSH30850.02EA 30.9 EUR
HYCLSH30850.02 HYCLSH30850.03
Dulbecco's phosphate buffered saline (DPBS), ES cell qualified, HyClone™ AdvanceSTEM™
Tampons Tampons de culture de tissus
A balanced salt solution tested for use with embryonic stem cells.
  • Without calcium and magnesium
  • 0,1 µm filter sterilised
  • Storage temperature of 15 to 30 °C
Complies with cGMP and ISO 91:2 guidelines.
Attention: For further manufacturing or research use. Not for diagnostic or therapeutic use.