Imprimer
Pierce™ Detergent Removal Spin Columns and Resin
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
87777 87778 87776 87779 87780
PIER87777EA 309 EUR
PIER87777 PIER87778 PIER87776 PIER87779 PIER87780
Pierce™ Detergent Removal Spin Columns and Resin
Protéines extraction Réactifs pour l'extraction des protéines
Pierce™ Detergent removal resin enables removal of commonly used ionic, non-ionic and zwitterionic detergents from protein/peptide digest solutions.
  • Fast and efficient protocol
  • Removes >95% of detergent from samples with high protein recovery
  • Improves mass spectrometry peptide coverage
  • Eliminates detergent-interference in downstream applications
The Pierce™ Detergent removal resin removes detergents with >95% efficiency at high concentrations (1 to 5%), while providing exceptionally high protein/peptide recovery for samples >100 µg/ml. For sample concentrations ≤100 µg/ml, Thermo Scientific™ HiPPR detergent removal resin (high protein and peptide recovery) is recommended.