Imprimer
L-Glutamine, HyClone™
Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30034.01 SH30034.02
HYCLSH30034.01EA 23.5 EUR
HYCLSH30034.01 SH30034.02
L-Glutamine, HyClone™
Suppléments et additifs pour culture cellulaire
HyClone™ L-glutamine is used as a supplement in serum- and protein-free media. It supports the growth of cells that have high energy demands and synthesise large amounts of proteins and nucleic acids.