Imprimer
Immobilised Streptavidin resin, UltraLink™, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
53114 53117 53113 53116
PIER53114EA 648 EUR
PIER53114 PIER53117 PIER53113 PIER53116
Immobilised Streptavidin resin, UltraLink™, Pierce™
Protéines purification Protein Purification Resins
Pierce™ Streptavidin UltraLink™ resin consists of purified recombinant streptavidin that has been covalently immobilised onto a beaded acrylamide support. UltraLink™ beads are hydrophilic, charge free, highly crosslinked, rigid and durable which makes the affinity resin suitable for a variety of batch or column type affinity procedures that require medium pressure chromatography.
  • Acrylamide based support provides an alternative to crosslinked beaded agarose supports for medium pressure applications
Streptavidin UltraLink™ resin binding capacity: >2 mg of biotinylated BSA/ml of resin
Streptavidin Plus UltraLink™ resin binding capacity: >4 mg of biotinylated BSA/ml of resin