Imprimer
HeLa protein digest standard, Pierce™
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
88328 88329
PIER88328EA 171 EUR
PIER88328 PIER88329
HeLa protein digest standard, Pierce™
Protein Purification Protein Purification Reagents
The HeLa Protein Digest Standard is a highly validated mammalian protein digest that may be used as a quality control sample for mass spectrometry (MS) analysis of complex proteomic samples.
  • Complex mammalian proteome sample protein digest (>15,000 proteins) from the HeLa S3 cell line
  • Prepared using both LysC and trypsin to ensure less than 10% tryptic missed cleavages
  • Validated peptide quality with <10% methionine oxidation and <10% lysine carbamylation
  • High-quality, consistent protein digestion backed up by rigorous testing and documentation
  • Provided in a stable lyophilised format