Imprimer
HyClone™ Ham's Nutrient Mixture F10/F12, Cell Culture Media, Powdered
Marques : HyClone products (Cytiva)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
SH30010.02 SH30010.03 SH30010.04
HYCLSH30010.02EA 61.3 EUR
HYCLSH30010.02 HYCLSH30010.03 HYCLSH30010.04
HyClone™ Ham's Nutrient Mixture F10/F12, Cell Culture Media, Powdered
Milieux de culture cellulaire
Dry powdered media for supporting cell culture growth and biomanufacturing processes.
  • Raw material components screened through strict quality control testing for lot-to-lot consistency.
  • Products are hydrated using purified process water and undergo sterile filtration.
  • Manufactured in cGMP (21 CFR 820) compliant and ISO9001 certified facilities.
Attention: Storage 2° to 8°C, away from light