Imprimer
Pierce™, EZ-Link™ Sulfhydryl Reactive Biotinylation Reagents
Marques : Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
21333 21341 21901BID 21900 21334 21911
PIER21333EA 289 EUR
PIER21333 PIER21341 PIER21901BID PIER21900 PIER21334 PIER21911
Pierce™, EZ-Link™ Sulfhydryl Reactive Biotinylation Reagents
Systèmes de liaison et de marquage des protéines
Sulfhydryl groups are the second most common target for modification in biological studies. They are found as exposed cysteines in protein and peptides. Sulfhydryl group biotinylation provides an advantage in some applications; for example, targeting a sulfhydryl group can help preserve the biological activity of an enzyme when amines are found at the active site.
Maleimide-PEG₂ and PEG₁₁-Biotin: Water-soluble. Reacts with sulfhydryl (-SH) groups at acidic to neutral pH. PEG₁₁-Biotin has an extra long spacer arm that reduces steric hindrance.
Biotin-BMCC: Reacts with sulfhydryl (-SH) groups at 6,5 to 7,5 pH. Avoids potential modification of tyrosine residues that can be associated with Iodoacetyl-Biotin. Must be dissolved in DMSO or DMF before use.
Iodoacetyl-PEG₂-Biotin: Water soluble. Reacts with sulfhydryl (-SH) groups at basic to neutral pH. Coupling reactions occur in the dark.
Iodoacetyl-LC-Biotin: Reacts with sulfhydryl (-SH) groups at basic to neutral pH. Coupling reactions occur in the dark. Must be dissolved in DMSO or DMF before use.
Biotin-HPDP: Pyridyldithiol reacts with sulfhydryl (-SH) groups to form a stable disulfide bond between pH 6 to 9. Pyridine-2-thione leaving group can be used to measure degree of biotinylation at 343 nm. Disulfide bond is cleavable using 50 mM DTT or 100 mM 2-mercaptoethanol. Must be dissolved in DMSO or DMF before use.