Imprimer
Pierce™ Immunoassay Microplates, Steptavidin Coated, 96-Well
Fournisseur: Thermo Fisher Scientific
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
15118 15124 15120 15122 15502 15119 15121 15125 15126 15500 15501 15503 15218 15219
PIER15118EA 306 EUR
PIER15118 PIER15124 PIER15120 PIER15122 PIER15502 PIER15119 PIER15121 PIER15125 PIER15126 PIER15500 PIER15501 PIER15503 PIER15218 PIER15219
Pierce™ Immunoassay Microplates, Steptavidin Coated, 96-Well
Microplaques Microplaques immunologiques et bandelettes
Pierce™ streptavidin coated PS microplates are ideal for binding assays using biotinylated antibodies. High binding capacity (HBC) plates provide a wider detection range and better curve linearity for small biotinylated ligands, such as peptides and oligonucleotides, than the standard coated plates. Available in clear for colorimetric assays, white for chemiluminescent assays, and black for fluorescent assays.
  • Low non specific binding
  • Easy and gentle immobilisation of biotin-containing conjugates
  • No denaturing of the protein component of a conjugate upon binding
  • Pre-blocked with Blocker™ BSA or SuperBlock™ blocking buffer and ready-to-use
  • High binding capacity plates provides wider detection range and better curve linearity for small biotinylated ligands
Biotin-binding plates are ideal for applications when direct adsorption to polystyrene denatures antibodies or the target molecule. Streptavidin is the most popular and widely available biotin-binding protein for research methods. Streptavidin has an isoelectric point of 5 to 6, resulting in low nonspecific interactions.