Imprimer
Ficoll® 400, molecular biology grade
Fournisseur: Merck Millipore (Calbiochem‎)
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
341691-25 341691-250
341691-25EA 363 EUR
341691-25 341691-250
Ficoll® 400, molecular biology grade
Milieux de séparation cellulaire
Ficoll® 400 is useful for the separation of lymphocyte sub populations. DNases, proteases, RNases: none detected.