Imprimer
Erlenmeyer flasks, disposable, with vented screw cap, Corning®
Fournisseur: Corning
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
11501 11502
CORN11501EA 796 EUR
CORN11501 CORN11502
Erlenmeyer flasks, disposable, with vented screw cap, Corning®
Ballons Flacons Erlenmeyers
PC, with PP vented screw cap, plain or baffled bottom.
  • Molded-in graduations for accuracy
  • Large vent cap for continuous gas exchange while ensuring sterility and preventing leakage
  • Sterility Assurance Level (SAL) of 10⁻⁶
  • Nonpyrogenic and RNase-/DNase-free
These Erlenmeyer flasks are designed to help scale-up suspension culture with space efficiency in mind. The flasks are sterile and ready-to-use, ideal for shaker culture applications, and storage. Autoclavable once at 121 °C for 20 minutes. Repeated autoclaving weakens the flask and is not recommended. Vent cap is not autoclavable.
Attention: *Le(s) produit(s) sur cette page sont hors du champ d'application de la directive européenne 2019/904