Imprimer
Powdered Trace Minerals for an Ultra-Clean Environment, MP Biomedicals
Fournisseur: MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
02960264.5 0296026401
ICNA02960264.5EA 135 EUR
ICNA02960264.5 ICNA0296026401
Powdered Trace Minerals for an Ultra-Clean Environment, MP Biomedicals
Fournitures pour animaux Alimentation animale
This animal diet supplement has seven-trace elements, which are usually present in a open environment - For complete description see product datasheet.
Trace Minerals is used in ultra-clean environments, where trace elements usually present, are absent.