Imprimer
Rose Bengal, poudre rouge foncé
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Sodium 4,5,6,7-tetrachloro-2',4',5',7'-tetraiodo-3-oxo-3H-spiro[isobenzofuran-1,9'-xanthene]-3',6'-bis(olate), Food Red No. 105, CI 45440, Acid red 94, Aniline Rose, Rose Bengal sodium
0219549125 0219549105
ICNA0219549125EA 124 EUR
ICNA0219549125 ICNA0219549105
Rose Bengal, poudre rouge foncé
Rose Bengal
Rose Bengal is a fluorescent probe for determining NAD binding sites of alcohol dehydrogenase.
Used in Conn′s technique for bacteria in soil; as an alternative to phloxine B in Kreyberg′s stain for keratin and mucus.
Used in Conn′s technique for bacteria in soil; as an alternative to phloxine B in Kreyberg′s stain for keratin and mucus.
Soluble in water at 1 mg/ml (pink to red solution).(Lit.)
Formule: C₂₀H₂Cl₄I₄O₅Na₂
Poids moléculaire: 1017.63 g/mol
Température de stockage: Température ambiante
Numéro MDL: MFCD00005043
Numéro CAS: 632-69-9
Commander maintenant
Résultats des tests de spécification
Identity Test Passes
UV/Vis: λmax (water) Store at Room Temperature (15 to 30 °C)