Imprimer
Ricinoleic acid ≥99%, liquide
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Ricinolic acid, (R)-12-Hydroxy-cis-9-octadecenoic acid, 12-Hydroxyoleic acid, Ricinelaidic acid, (R,Z)-12-Hydroxyoctadec-9-enoic acid
0219151490 0219151480
ICNA0219151490EA 293 EUR
ICNA0219151490 ICNA0219151480
Ricinoleic acid ≥99%, liquide
Ricinoleic acid
Ricinoleic Acid is a naturally occurring 12-hydroxy fatty acid constituting about 90% of the fatty acids in castor oil. Has the capacity to serve as a substrate for the synthesis of the conjugated linoleic acids.
Ricinoleic Acid is used as a substrate for the synthesis of the conjugated linoleic acids.
Formule: C₁₈H₃₄O₃
Poids moléculaire: 298.47 g/mol
Numéro MDL: MFCD00084840
Numéro CAS: 141-22-0