Imprimer

2-(4-Isobutylphenyl)propanoic acid, en poudre

Marques : MP Biomedicals
Ratings: Ecrire un avis

Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5

Attention

0219021601 0219021605
ICNA0219021601EA 80.1 EUR
ICNA0219021601 ICNA0219021605
2-(4-Isobutylphenyl)propanoic acid, en poudre
2-(4-Isobutylphenyl)propanoic acid

Ibuprofen an anti-inflammatory and analgesic.


Ibuprofen is clinically useful as a nonsteroidal anti-inflammatory drug.


Ibuprofen is clinically useful as a nonsteroidal anti-inflammatory drug.


Cyclooxygenase (COX) inhibitor that has greater activity against COX-1 than against COX-2. Also inhibits cyclooxygenase, PGH synthase-1 and PGH synthase-2. Ibuprofen is reported to inhibit the netrophil-dependent edema response of rabbit skin to C5a/PGE23.


Freely soluble in acetone; dichloromethane, ethanol; soluble in methanol (at least 50 mg/mL-clear, colorless solution) and most organic solvents; Insoluble in water.


Attention: Research Use Only (RUO).

Formule: C₁₃H₁₈O₂
Température de stockage: Température ambiante
Numéro MDL: MFCD00010393
Numéro CAS: 15687-27-1

Commander maintenant

Résultats des tests de spécification

Identity Test Passes
FTIR Conforms to Standard
Melting Point 72 - 82 °C


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Plus About VWR