Imprimer
Peptone, powder, bacteriological grade, MP Biomedicals
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
0210480801 0210480805
ICNA0210480801EA 228 EUR
ICNA0210480801 ICNA0210480805
Peptone, powder, bacteriological grade, MP Biomedicals
Milieux pour microbiologie Matières premières de microbiologie
Soluble in water (100 mg/ml - clear to slightly hazy solution).
  • Aerobic Plate Count ≤1000 cfu/g
  • Yeasts/Molds <100 cfu/g; Nitrogen ≥14%
Peptone is used in culture media in bacteriology, the growth requirements of most non fastidious organisms will be fulfilled by the range of amino acids, peptides and proteoses in this mixture.