Imprimer
Taurocholic acid sodium salt ≥97%, en poudre, "Practical"
Marques : MP Biomedicals
Ratings: Ecrire un avis
Total Evaluation: 0
Moyenne des Notes : 0.0 sur 5
Synonyme(s): Cholaic acid sodium salt, Taurocholic acid sodium, Sodium 2-((R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanamido)ethane-1-sulphonate (1:1), Sodium taurocholate
0210293080 0210293090 0210293025
ICNA0210293080EA 2746 EUR
ICNA0210293080 ICNA0210293090 ICNA0210293025
Taurocholic acid sodium salt ≥97%, en poudre, "Practical"
Taurocholic acid sodium salt
Applications
Taurocholic Acid Sodium Salt is an anionic detergent used for protein solubilization.
Product Description
Taurocholic Acid Sodium Salt is the conjugation product of cholic acid with taurine and the principal constituent of the bile of carnivorous animals.
Biochem/physiol Actions
Non-sulfated bile salt, physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp), Na(+)/taurocholate cotransporter (NTCP) and MRP3 into the hepatic carnalicular.
Melting Point: 230 °C (decomposes)(Lit.)
Presentation: White Powder
Format: Powder
pH: 6 - 7.5(1% aq soln)
Aggregation Number: 4
Critical Micelle Concentration (CMC): 3-11 mM
Heavy Metals: =20 ppm
Loss on Drying: =7.0%
NOTES: Sodium Cholate : =3.0%
Solubility: Soluble in water (100 mg/mL).
Storage & Handling: Store at Room Temperature (15-30 °C).
Taurocholic Acid Sodium Salt is the conjugation product of cholic acid with taurine and the principal constituent of the bile of carnivorous animals.
Taurocholic Acid Sodium Salt is an anionic detergent used for protein solubilisation.
Non-Sulphated bile salt, physiological transport substrate for the bile salt export pump/sister of Pgp (BSEP/spgp), Na(+)/taurocholate cotransporter (NTCP) and MRP3 into the hepatic carnalicular.
Formule: C₂₆H₄₄NNaO₇S
Poids moléculaire: 537.688 g/mol
Numéro MDL: MFCD00013050
Numéro CAS: 145-42-6